missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-1,4-benzoquinone, TRC

Product Code. 30135218
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135218 5 g 5g
30135077 1 g 1g
30008335 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135218 Supplier Toronto Research Chemicals Supplier No. TRCD4320255G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697-91-6
Formula Weight 175.943
InChI Formula InChI=1S/C6H2Cl2O2/c7-4-1-3(9)2-5(8)6(4)10/h1-2H
IUPAC Name 2,6-dichlorocyclohexa-2,5-diene-1,4-dione
Molecular Formula C6 H2 Cl2 O2
Molecular Weight (g/mol) 176.985
SMILES ClC1=CC(=O)C=C(Cl)C1=O
Synonym 2,6-Dichloro-2,5-cyclohexadiene-1,4-dione, 2,6-Dichloro-p-benzoquinone, 2,6-Dichlorobenzoquinone, 2,6-Dichloroquinone, NSC 6211;
Chemical Name or Material 2,6-Dichloro-1,4-benzoquinone

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.