missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-1,4-benzoquinone, 97+%

Product Code. p-7030016
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461830 5 g 5g
11471830 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461830 Supplier Thermo Scientific Alfa Aesar Supplier No. H27542.06
Add to basket
This item is not returnable. View return policy

CAS: 697-91-6 | C6H2Cl2O2 | 176.98 g/mol

2,6-Dichloro-1,4-benzoquinone is the suitable reagent used to in a study to evaluate the diffusion coefficients (D) for a family of quinones, nitroaromatics, ferrocenes and aromatic hydrocarbon compounds, in acetonitrile by single potential step chronoamperometry.1 It may be used in the preparation of 2,3,5-Trichloro-1,4-dihydroquinone, 2,3,5-trichloro-1,4-benzoquinone and 2,6-Dichloro-1,4-dihydroquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloro-1,4-benzoquinone is the suitable reagent used to in a study to evaluate the diffusion coefficients (D) for a family of quinones, nitroaromatics, ferrocenes and aromatic hydrocarbon compounds, in acetonitrile by single potential step chronoamperometry.1 It may be used in the preparation of 2,3,5-Trichloro-1,4-dihydroquinone, 2,3,5-trichloro-1,4-benzoquinone and 2,6-Dichloro-1,4-dihydroquinone.

Solubility
Insoluble in water. Solubility in methanol (almost transparency).

Notes
Store away from reducing agents and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697-91-6
Molecular Formula C6H2Cl2O2
Molecular Weight (g/mol) 176.98
MDL Number MFCD00037159
InChI Key JCARTGJGWCGSSU-UHFFFAOYSA-N
Synonym 2,6-dichloro-1,4-benzoquinone, 2,6-dichloro-p-benzoquinone, 2,6-dichlorobenzoquinone, 2,6-dichloroquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-dichloro, 2,6-dichloquinone, p-quinone, 2,6-dichloro, p-benzoquinone, 2,6-dichloro, 2,6-dichloro-2,5-cyclohexadiene-1,4-dione, p-quinone,6-dichloro
PubChem CID 12771
IUPAC Name 2,6-dichlorocyclohexa-2,5-diene-1,4-dione
SMILES C1=C(C(=O)C(=CC1=O)Cl)Cl

Specifications

Melting Point 119°C to 121°C
Quantity 5 g
Solubility Information Insoluble in water. Solubility in methanol (almost transparency).
Formula Weight 176.98
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-1,4-benzoquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.