Learn More
2,6-Dibromonaphthalene-1,5-diol, 93%, BLD Pharm™
Specifications
Specifications
| CAS | 84-59-3 |
| Molecular Formula | C10H6Br2O2 |
| MDL Number | MFCD00021473 |
| Quantity | 25 g |
| SMILES | OC1=C2C=CC(Br)=C(O)C2=CC=C1Br |
| Molecular Weight (g/mol) | 317.96 |
| Percent Purity | 93% |
| Chemical Name or Material | 2,6-Dibromonaphthalene-1,5-diol |