Learn More
2,6-Dibromo-4-(trifluoromethoxy)benzenamine, TRC
Product Code. 30136504
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136504 50 g 50g
30010314 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30136504 Supplier Toronto Research Chemicals Supplier No. TRCD42620050G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 88149-49-9
Molecular Formula C7H4Br2F3NO
Molecular Weight (g/mol) 334.92
Synonym 2,6-Dibromo-4-trifluoromethoxyaniline
SMILES BrC1=C(N)C(Br)=CC(OC(F)(F)F)=C1
Specifications
Quantity 50 g
Chemical Name or Material 2,6-Dibromo-4-(trifluoromethoxy)benzenamine