Learn More
2,6-Di-tert-butyl-p-benzoquinone, TRC
Product Code. 30006878
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006878 10 g 10g
30006743 25 g 25g
30003493 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006878 Supplier Toronto Research Chemicals Supplier No. TRCD42943510G
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 719-22-2
Molecular Formula C14 H20 O2
Molecular Weight (g/mol) 220.31
Synonym 2,6-Bis(1,1-dimethylethyl)-2,5-cyclohexadiene-1,4-dione, 2,6-Bis[1,1-dimethyl ethyl]quinone, 2,6-Di-tert-butyl-1,4-benzoquinone, 2,6-Di-tert-butyl-2,5-cyclohexadien-1,4-dione, 2,6-Di-tert-butyl-2,5-cyclohexadiene-1,4-dione, 2,6-Di-tert-butyl-p-benzoquinone, 2,6-Di-tert-butyl-p-quinone, 2,6-Di-tert-butylbenzoquinone, 2,6-Di-tert-butylquinone, 3,5-Di-tert-butyl-p-benzoquinone, 3,5-Di-tert-butylquinone, NSC 14448
IUPAC Name 2,6-ditert-butylcyclohexa-2,5-diene-1,4-dione
SMILES CC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C
Specifications
Quantity 10 g
InChI Formula InChI=1S/C14H20O2/c1-13(2,3)10-7-9(15)8-11(12(10)16)14(4,5)6/h7-8H,1-6H3
Formula Weight 220.1463
Chemical Name or Material 2,6-Di-tert-butyl-p-benzoquinone