Learn More
2,6-Bis(1,1-dimethylethyl)-4-(4-morpholinylmethyl)phenol, TRC
Specifications
Specifications
| Chemical Name or Material | 2,6-Bis(1,1-dimethylethyl)-4-(4-morpholinylmethyl)phenol |
| CAS | 2773-50-4 |
| Molecular Formula | C19 H31 N O2 |
| Quantity | 50 mg |
| Synonym | 2,6-Di-tert-butyl-4-(morpholinomethyl)phenol |
| InChI Formula | InChI=1S/C19H31NO2/c1-18(2,3)15-11-14(13-20-7-9-22-10-8-20)12-16(17(15)21)19(4,5)6/h11-12,21H,7-10,13H2,1-6H3 |
| SMILES | OC=1C(=CC(=CC1C(C)(C)C)CN2CCOCC2)C(C)(C)C |
| IUPAC Name | 2,6-di-tert-butyl-4-(morpholinomethyl)phenol |
| Molecular Weight (g/mol) | 305.46 |
| Formula Weight | 305.24 |