Learn More
2-(6-Benzyloxyindolyl)ethylamine Hemisulfate, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(6-Benzyloxyindolyl)ethylamine Hemisulfate |
| CAS | 31677-74-4 |
| Molecular Formula | 2 C17 H18 N2 O . H2 S O4 |
| Quantity | 10 mg |
| Synonym | 3-(2-Aminoethyl)-6-(benzyloxy)indole Hemisulfate |
| InChI Formula | InChI=1S/2C17H18N2O.H2O4S/c2*18-9-8-14-11-19-17-10-15(6-7-16(14)17)20-12-13-4-2-1-3-5-13;1-5(2,3)4/h2*1-7,10-11,19H,8-9,12,18H2;(H2,1,2,3,4) |
| SMILES | NCCc1c[nH]c2cc(OCc3ccccc3)ccc12.NCCc4c[nH]c5cc(OCc6ccccc6)ccc45.OS(=O)(=O)O |
| IUPAC Name | 2-(6-phenylmethoxy-1H-indol-3-yl)ethanamine;sulfuric acid |
| Molecular Weight (g/mol) | 630.754 |
| Formula Weight | 630.251 |