missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N,N-bis(2-pyridylethyl)amine Trihydrochloride, TRC

Product Code. 17904225
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904225 2.5 g 2.5g
17955095 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17904225 Supplier Toronto Research Chemicals Supplier No. TRCM2952552.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2095467-43-7
InChI Formula InChI=1S/C15H19N3.3ClH/c1-18(12-8-14-6-2-4-10-16-14)13-9-15-7-3-5-11-17-15;;;/h2-7,10-11H,8-9,12-13H2,1H3;3*1H
IUPAC Name N-methyl-2-pyridin-2-yl-N-(2-pyridin-2-ylethyl)ethanamine;trihydrochloride
Molecular Formula C15 H19 N3 . 3 Cl H
Molecular Weight (g/mol) 350.71
Chemical Name or Material N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridin-2-yl)ethyl]ethanamine Trihydrochloride

Specifications

Quantity 2.5 g
Synonym 2-Pyridineethanamine, N-methyl-N-[2-(2-pyridinyl)ethyl]-, hydrochloride (1:3), N-Methyl-N-[2-(2-pyridinyl)ethyl]-2-pyridineethanamine trihydrochloride, Methylbis(2-pyridylethyl)amine trihydrochloride, N-Methylbis[2-(2-pyridylethyl)]amine trihydrochloride, N-Methyl-N,N-bis[2-(2-pyridylethyl)]amine trihydrochloride, N-Methyl-2-(pyridin-2-yl)-N-[2-(pyridin-2-yl)ethyl]ethanamine trihydrochloride
SMILES Cl.Cl.Cl.CN(CCc1ccccn1)CCc2ccccn2
Formula Weight 349.0879
Percent Purity >95%
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.