Learn More
2-(5-formylfuran-2-yl)benzoic Acid, TRC
Product Code. 30407723
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407723 10 mg
30395873 100 mg
30393595 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30407723 Supplier Toronto Research Chemicals Supplier No. TRCF68502510MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 88460-72-4
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.18
SMILES OC(=O)c1ccccc1c2oc(C=O)cc2
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C12H8O4/c13-7-8-5-6-11(16-8)9-3-1-2-4-10(9)12(14)15/h1-7H,(H,14,15)
Formula Weight 216.0423
Chemical Name or Material 2-(5-formylfuran-2-yl)benzoic Acid