missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-4-(4-morpholinylmethyl)phenol hydrochloride hydrate, 99%, Thermo Scientific™

Product Code. 11356756
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356756 100 g 100g
11346756 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356756 Supplier Thermo Scientific Alfa Aesar Supplier No. L04024.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33625-43-3
Molecular Formula C13H20ClNO2
Molecular Weight (g/mol) 257.76
MDL Number MFCD00012796
InChI Key RLGUYDSHQWRANT-UHFFFAOYSA-N
Synonym 2,5-dimethyl-4-morpholinomethyl phenol hydrochloride, phenol, 2,5-dimethyl-4-4-morpholinylmethyl-, hydrochloride, phenol, 2,5-dimethyl-4-4-morpholinylmethyl-, hydrochloride 1:1, 2,5-dimethyl-4-morpholin-4-ylmethyl phenol hydrochloride, 2,5-dimethyl-4-morpholin-4-ylmethyl phenol, chloride, 4-morpholinomethyl-2,5-xylenol hydrochloride, acmc-20aoah, 2,5-dimethyl-4-morpholinomethyl phenolhydrochloride, 2,5-dimethyl-4-4-morpholinylmethyl phenol hydrochloride hydrate
PubChem CID 118512
IUPAC Name 2,5-dimethyl-4-(morpholin-4-ylmethyl)phenol;hydrochloride
SMILES Cl.CC1=CC(CN2CCOCC2)=C(C)C=C1O

Specifications

Melting Point 181°C to 185°C
Quantity 100 g
Beilstein 4024300
Formula Weight 257.76 (Anhydrous)
Percent Purity 99%
Chemical Name or Material 2,5-Dimethyl-4-(4-morpholinylmethyl)phenol hydrochloride hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.