Learn More
2,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-pyridine (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-pyridine |
| CAS | 1309443-99-9 |
| Molecular Formula | C13H20BNO2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C13H20BNO2/c1-9-8-15-10(2)7-11(9)14-16-12(3,4)13(5,6)17-14/h7-8H,1-6H3 |
| SMILES | Cc1cc(B2OC(C)(C)C(C)(C)O2)c(C)cn1 |
| IUPAC Name | 2,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
| Molecular Weight (g/mol) | 233.11 |
| Formula Weight | 233.1587 |