Learn More
2,5-Dimethoxyphenethylamine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Dimethoxyphenylethylamine Hydrochloride |
| CAS | 3166-74-3 |
| Molecular Formula | C10 H15 N O2 . Cl H |
| Quantity | 10 mg |
| Synonym | 2C-H (hydrochloride), 2,5-Dimethoxyphenylethylamine HCl |
| InChI Formula | InChI=1S/C10H15NO2.ClH/c1-12-9-3-4-10(13-2)8(7-9)5-6-11;/h3-4,7H,5-6,11H2,1-2H3;1H |
| SMILES | Cl.COc1ccc(OC)c(CCN)c1 |
| IUPAC Name | 2-(2,5-dimethoxyphenyl)ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 217.692 |
| Formula Weight | 217.087 |