Learn More
2,5-Dimethoxy-p-toluic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Dimethoxy-p-toluic Acid |
| CAS | 32176-94-6 |
| Molecular Formula | C10H12O4 |
| Quantity | 10 mg |
| Synonym | 2,5-Dimethoxy-4-methylbenzoic Acid, 4-Methyl-2,5-dimethoxybenzoic Acid |
| InChI Formula | InChI=1S/C10H12O4/c1-6-4-9(14-3)7(10(11)12)5-8(6)13-2/h4-5H,1-3H3,(H,11,12) |
| SMILES | COc1cc(C(=O)O)c(OC)cc1C |
| IUPAC Name | 2,5-dimethoxy-4-methylbenzoic acid |
| Molecular Weight (g/mol) | 196.2 |
| Formula Weight | 196.0736 |