missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorotoluene, 98%, Thermo Scientific™

Product Code. 11475107
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11475107

Brand: Thermo Scientific Alfa Aesar B22544.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-67-5
Molecular Formula C7H6F2
Molecular Weight (g/mol) 128.12
MDL Number MFCD00000347
InChI Key YSNVKDGEALPJGC-UHFFFAOYSA-N
Synonym 2,5-difluorotoluene, benzene, 1,4-difluoro-2-methyl, 1,4-difluoro-2-methyl-benzene, 1,4-difluor-2-methylbenzol, 2,5-difluorotolune, 2,5-difluoro toluene, p-ditrifluorotoluene, acmc-209k1g, ksc236k9r, benzene,4-difluoro-2-methyl
PubChem CID 67980
IUPAC Name 1,4-difluoro-2-methylbenzene
SMILES CC1=CC(F)=CC=C1F

Specifications

Melting Point -35°C
Density 1.36
Boiling Point 115°C to 117°C
Flash Point 12°C (53°F)
Odor Aromatic
Refractive Index 1.453
Quantity 25 g
UN Number UN2388
Beilstein 2041492
Formula Weight 128.12
Percent Purity 98%
Chemical Name or Material 2,5-Difluorotoluene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.