Learn More
2,5-Difluorobenzoic-d3 Acid, CDN
Product Code. 30328401
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328401 0.25 g 0.25g
30315663 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30328401 Supplier CDN Supplier No. CDND64840.25G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,5-Difluorobenzoic Acid-D3
CAS 1219798-63-6
Molecular Formula C7 D3 H F2 O2
Quantity 0.25 g
Synonym 2,5-Difluorobenzoic-d3 Acid, 2,5-Difluorobenzoic Acid-D3, 3,6-Difluorobenzoic-2,4,5-d3 Acid
InChI Formula InChI=1 S/C7H4F2O2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3 H,(H,10,11)/i1D,2 D,3 D
SMILES [2 H]c1c([2 H])c(F)c(C(=O)O)c([2 H])c1F
IUPAC Name 2,4,5-trideuterio-3,6-difluorobenzoic acid
Molecular Weight (g/mol) 161.1208
Formula Weight 161.0368 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity