missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Product Code. 15496247
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496247 250 mg 250mg
15486247 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496247 Supplier Thermo Scientific Alfa Aesar Supplier No. H33442.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89719-92-6 | Molecular Formula: C6H2Cl2F3N | Molecular Weight: 215.98 g/mol

2-chloro-6-(trifluoromethyl)pyridine, 2-chloro-5-(trifluoromethyl)pyridine and 3-chloro-4-(trifluoromethyl)pyridine were each converted into the three possible carboxylic acids 2, 4, 6,8, 10, 12, 16, 17 and 20. This was achieved by employing several, but not all of the organometallic toolbox methods : transformation of a more basic organometallic species into a less basic isomer by transmetalation-equilibration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-chloro-6-(trifluoromethyl)pyridine, 2-chloro-5-(trifluoromethyl)pyridine and 3-chloro-4-(trifluoromethyl)pyridine were each converted into the three possible carboxylic acids 2, 4, 6,8, 10, 12, 16, 17 and 20. This was achieved by employing several, but not all of the organometallic “toolbox methods”: transformation of a more basic organometallic species into a less basic isomer by transmetalation-equilibration.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, aluminum. Protection under nitrogen and lower temperature is recommended(below 4 deg C).

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89719-92-6
Molecular Formula C6H2Cl2F3N
Molecular Weight (g/mol) 215.98
MDL Number MFCD09749663
InChI Key GFTRYJQAASFSIB-UHFFFAOYSA-N
Synonym 2,5-dichloro-4-trifluoromethyl pyridine, pyridine, 2,5-dichloro-4-trifluoromethyl, acmc-20dwro
PubChem CID 26986016
IUPAC Name 2,5-dichloro-4-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC(Cl)=NC=C1Cl

Specifications

Boiling Point 76°C (18 mmHg)
Quantity 250 mg
UN Number UN1993
Formula Weight 215.99
Percent Purity 97%
Chemical Name or Material 2,5-Dichloro-4-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.