missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diacetoxytoluene, 97%, Thermo Scientific™

Product Code. 11406396
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11406396

Brand: Thermo Scientific Alfa Aesar B23156.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 717-27-1
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00061098
InChI Key KUZVIVNLNXNLAQ-UHFFFAOYSA-N
Synonym 2,5-diacetoxytoluene, methylhydroquinone diacetate, 1,4-diacetoxy-2-methylbenzene, 1,4-benzenediol,2-methyl-, 1,4-diacetate, 4-acetyloxy-3-methylphenyl acetate, acmc-209ojx, methylhydroquinone, diacetate, 2-methylhydroquinone diacetate, paragos 310054
PubChem CID 4184462
IUPAC Name (4-acetyloxy-3-methylphenyl) acetate
SMILES CC1=C(C=CC(=C1)OC(=O)C)OC(=O)C

Specifications

Melting Point 40°C to 42°C
Quantity 100 g
Beilstein 1967373
Formula Weight 208.22
Percent Purity 97%
Chemical Name or Material 2,5-Diacetoxytoluene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.