Learn More
2,5-Bis(trifluoromethyl)benzoic acid, 98%
Product Code. 11366579
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366579 1 g 1g
11376579 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11366579 Supplier Thermo Scientific Alfa Aesar Supplier No. H26184.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 42580-42-7 | C9H4F6O2 | 258.119 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42580-42-7
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.119
MDL Number MFCD00013249
InChI Key PINBPLCVZSKLTF-UHFFFAOYSA-N
PubChem CID 736187
ChEBI CHEBI:60697
IUPAC Name 2,5-bis(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1C(F)(F)F)C(=O)O)C(F)(F)F
Specifications
Melting Point 77°C to 80°C
Quantity 1 g
Synonym 2,5-bis trifluoromethyl benzoic acid, 2,5-di trifluoromethyl benzoic acid, 2,5-bis trifluoroformyl benzoic acid, pubchem8492, acmc-1amwk, rarechem al bo 1002, 2,5-bis-trifluoromethyl-benzoic acid, 2,5-bis trifluoromethyl benzoic acid., benzoic acid,2,5-bis trifluoromethyl, benzoic acid, 2,5-bis trifluoromethyl
Formula Weight 258.12
Percent Purity 98%
Chemical Name or Material 2,5-Bis(trifluoromethyl)benzoic acid