missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Bis(trifluoromethyl)acetophenone, 97%, Thermo Scientific™

Product Code. 11342569
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11342569

Brand: Thermo Scientific Alfa Aesar H26659.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Bis(trifluoromethyl)acetophenone is used in the synthesis of the monoamine oxidase-B inhibitors. It is also used to isolate novel bacterial strain from soil samples.

Solubility
Sparingly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 545410-47-7
Molecular Formula C10H6F6O
Molecular Weight (g/mol) 256.147
MDL Number MFCD04116044
InChI Key ZBIHDCGAPRWDOA-UHFFFAOYSA-N
Synonym 2',5'-bis trifluoromethyl acetophenone, 1-2,5-bis trifluoromethyl phenyl ethanone, 2,5-bis trifluoromethyl acetophenone, 1-2,5-bis-trifluoromethyl-phenyl-ethanone, ethanone, 1-2,5-bis trifluoromethyl phenyl, 2,5-bis-trifluoromethyl acetophenone, 1-acetyl-2,5-bis trifluoromethyl benzene, 2,5-bistrifluoromethyl-1-methylcarbonylbenzene, 1-2'-azido-2'-deoxy-, a-d-ribofuranosyl thymine
PubChem CID 3751362
IUPAC Name 1-[2,5-bis(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=C(C=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Refractive Index 1.42
Quantity 10 g
Solubility Information Sparingly soluble in water.
Formula Weight 256.15
Percent Purity 97%
Chemical Name or Material 2',5'-Bis(trifluoromethyl)acetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.