Learn More
2,5-Bis(phenylamino)-2,5-cyclohexadiene-1,4-dione, TRC
Product Code. 30347839
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347839 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30347839 Supplier Toronto Research Chemicals Supplier No. TRCB674595250MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,5-Bis(phenylamino)-2,5-cyclohexadiene-1,4-dione
CAS 3421-08-7
Molecular Formula C18 H14 N2 O2
Quantity 250 mg
Synonym 2,5-Bis(anilino)-1,4-benzoquinone, 2,5-Dianilino-1,4-benzoquinone, 2,5-Dianilino-p-benzoquinone, C.I. 56000, Helindon Yellow CA, Helindon Yellow CAK, NSC 11397, NSC 46456
InChI Formula InChI=1S/C18H14N2O2/c21-17-12-16(20-14-9-5-2-6-10-14)18(22)11-15(17)19-13-7-3-1-4-8-13/h1-12,19-20H
SMILES O=C1C=C(Nc2ccccc2)C(=O)C=C1Nc3ccccc3
IUPAC Name 2,5-dianilinocyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol) 290.316
Formula Weight 290.106