Learn More
2,5-Anhydro-D-glucitol, TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Anhydro-D-glucitol |
| CAS | 27826-73-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6 H12 O5 |
| Quantity | 25 mg |
| Synonym | D-Glucitol, 2,5-anhydro-, Glucitol, 2,5-anhydro-, D- (8CI), 2,5-Anhydro-D-glucitol, 2,5-Anhydro-D-sorbitol, 2,5-Anhydrosorbitol, 2,5-Sorbitan, (2R,3S,4S,5S)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran |
| InChI Formula | InChI=1S/C6H12O5/c7-1-3-5(9)6(10)4(2-8)11-3/h3-10H,1-2H2/t3-,4+,5-,6-/m1/s1 |
| SMILES | OC[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O |
| IUPAC Name | (2R,3S,4S,5S)-2,5-bis(hydroxymethyl)oxolane-3,4-diol |
| Molecular Weight (g/mol) | 164.16 |
| Show More |