Learn More
2,4-Nonanedione, 98+%
Product Code. 11344484
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11344484
Brand: Thermo Scientific Alfa Aesar L01151.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 6175-23-1 | C9H16O2 | 156.225 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6175-23-1
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00015276
InChI Key KFBXUKHERGLHLG-UHFFFAOYSA-N
Synonym 2,4-nonanedione, caproylacetone, 2,4-nonadione, acmc-20aobv
PubChem CID 80314
IUPAC Name nonane-2,4-dione
SMILES CCCCCC(=O)CC(=O)C
Specifications
Boiling Point 100°C to 103°C (19 mmHg)
Refractive Index 1.4608
Quantity 25 g
Beilstein 1757688
Formula Weight 156.23
Percent Purity ≥98%
Chemical Name or Material 2,4-Nonanedione