missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Morpholinyl)ethyl isothiocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11368585
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368585 10 g 10g
11358585 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368585 Supplier Thermo Scientific Alfa Aesar Supplier No. L12046.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.

Solubility
Hydrolyzes in water.

Notes
Store at 4°C. Store under dry inert gas. Protect from humidity and water. Incompatible with moisture, amines, alcohols and oxidizing agents. Protect from heat. Moisture Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63224-35-1
Molecular Formula C7H12N2OS
Molecular Weight (g/mol) 172.25
MDL Number MFCD00041221
InChI Key SDNSCXCXVQHBGH-UHFFFAOYSA-N
Synonym 2-morpholinoethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine, 2-4-morpholino ethyl isothiocyanate, 2-morpholinoethylisothiocyanate, 4-2-isothiocyanato-ethyl-morpholine, 2-morpholin-4-ylethanisothiocyanate, 2-4-morpholinyl ethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine #
PubChem CID 143957
IUPAC Name 4-(2-isothiocyanatoethyl)morpholine
SMILES S=C=NCCN1CCOCC1

Specifications

Boiling Point 162°C to 163°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN2922
Beilstein 1210530
Formula Weight 172.25
Percent Purity 97%
Chemical Name or Material 2-(4-Morpholinyl)ethyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.