missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylphenyl)propane-d14, TRC

Product Code. 30365422
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365422 50 mg 50mg
30359672 10 mg -
30360144 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30365422 Supplier Toronto Research Chemicals Supplier No. TRCC99410650MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93952-03-5
Molecular Formula C10 2H14
Molecular Weight (g/mol) 148.3
Synonym Benzene-1,2,4,5-d4, 3-(methyl-d3)-6-[1-(methyl-d3)ethyl-1,2,2,2-d4]- (9CI), 2-(4-Methylphenyl)propane-d14, 4-Isopropyltoluene D14, p-Cymene D14
IUPAC Name 1,2,4,5-tetradeuterio-3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-6-(trideuteriomethyl)benzene
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1C([2H])([2H])[2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H14/c1-8(2)10-6-4-9(3)5-7-10/h4-8H,1-3H3/i1D3,2D3,3D3,4D,5D,6D,7D,8D
Formula Weight 148.1974
Chemical Name or Material 4-Isopropyltoluene-d14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.