Learn More
2,4-Dithiouridine 2',3',5'-Triacetate, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Dithiouridine 2',3',5'-Triacetate |
| CAS | 28542-32-7 |
| Molecular Formula | C15H18N2O7S2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C15H18N2O7S2/c1-7(18)21-6-10-12(22-8(2)19)13(23-9(3)20)14(24-10)17-5-4-11(25)16-15(17)26/h4-5,10,12-14H,6H2,1-3H3,(H,16,25,26)/t10-,12-,13-,14-/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)N2C=CC(=S)NC2=S |
| IUPAC Name | [(2R,3R,4R,5R)-3,4-diacetyloxy-5-[2,4-bis(sulfanylidene)pyrimidin-1-yl]oxolan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 402.44 |
| Formula Weight | 402.0555 |