missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-3-methylbenzoyl chloride, 98%, Thermo Scientific™

Product Code. 15454267
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454267 250 mg 250mg
15444267 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454267 Supplier Thermo Scientific Alfa Aesar Supplier No. H31658.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112857-70-2
Molecular Formula C8H5ClF2O
Molecular Weight (g/mol) 190.574
MDL Number MFCD07777157
InChI Key BMMDDDOEASRDKX-UHFFFAOYSA-N
Synonym benzoyl chloride,2,4-difluoro-3-methyl, acmc-20mh3r, 2,4-difluoro-3-methyl-benzoyl chloride, 2,4-bis fluoranyl-3-methyl-benzoyl chloride, benzoyl chloride, 2,4-difluoro-3-methyl-9ci
PubChem CID 12137487
IUPAC Name 2,4-difluoro-3-methylbenzoyl chloride
SMILES CC1=C(C=CC(=C1F)C(=O)Cl)F

Specifications

Melting Point 35°C to 38°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 190.58
Percent Purity 98%
Chemical Name or Material 2,4-Difluoro-3-methylbenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.