Learn More
2,4'-Dichlorobenzophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4'-Dichlorobenzophenone |
| CAS | 85-29-0 |
| Molecular Formula | C13 H8 Cl2 O |
| Quantity | 10 g |
| Synonym | Benzophenone, 2,4'-dichloro- (6CI,7CI,8CI), (2-Chlorophenyl)(4-chlorophenyl)methanone, 2,4'-Dichlorobenzophenone, NSC 3221, o,p'-DCBP, o,p'-Dichlorobenzophenone |
| InChI Formula | InChI=1S/C13H8Cl2O/c14-10-7-5-9(6-8-10)13(16)11-3-1-2-4-12(11)15/h1-8H |
| SMILES | Clc1ccc(cc1)C(=O)c2ccccc2Cl |
| IUPAC Name | (2-chlorophenyl)-(4-chlorophenyl)methanone |
| Molecular Weight (g/mol) | 251.11 |
| Formula Weight | 249.9952 |