missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-1-fluorobenzene, 98%, Thermo Scientific™

Product Code. p-3211508 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10200643

Brand: Thermo Scientific Acros 380700050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1435-53-6
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.90
MDL Number MFCD00000283
InChI Key MKHDDTWHDFVYDQ-UHFFFAOYSA-N
Synonym 2,4-dibromofluorobenzene, 1,3-dibromo-4-fluorobenzene, 1-fluoro-2,4-dibromobenzene, benzene, 2,4-dibromo-1-fluoro, pubchem3446, acmc-209cqz, 1,3-dibromo-5-fluoro, intermediates-zcf02097, ksc494g9j
PubChem CID 15034
IUPAC Name 2,4-dibromo-1-fluorobenzene
SMILES FC1=CC=C(Br)C=C1Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 2.0470g/mL
Boiling Point 105°C to 110°C (22.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Br2C6H3F
Refractive Index 1.5820 to 1.584
Quantity 5 mL
Specific Gravity 2.047
Solubility Information Solubility in water: insoluble
Formula Weight 253.9
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Dibromo-1-fluorobenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.