missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-undecyl-1,3,5-triazine, 97%, Thermo Scientific™

Product Code. 11493987
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493987 25 g 25g
11473987 1 g 1g
11483987 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493987 Supplier Thermo Scientific Alfa Aesar Supplier No. A19392.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2533-34-8
Molecular Formula C14H27N5
Molecular Weight (g/mol) 265.405
MDL Number MFCD00213901
InChI Key MLCIKWISJBFZKS-UHFFFAOYSA-N
PubChem CID 75673
IUPAC Name 6-undecyl-1,3,5-triazine-2,4-diamine
SMILES CCCCCCCCCCCC1=NC(=NC(=N1)N)N

Specifications

Melting Point 119°C to 121°C
Quantity 25 g
Synonym lauroguanamine, 2,4-diamino-6-undecyl-s-triazine, s-triazine, 2,4-diamino-6-undecyl, 2,4-diamino-6-undecyl-1,3,5-triazine, acmc-1cdkx, 1,3,5-triazine-2,4-diamine, 6-undecyl, 4-26-00-01233 beilstein handbook reference, 1,3,5-triazine-2,4-diamine,6-undecyl, 1,3,5-triazine-2,4-diamine, 6-undecyl-9ci
Formula Weight 265.4
Percent Purity 97%
Chemical Name or Material 2,4-Diamino-6-undecyl-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.