Learn More
2-(4-Chlorophenoxy)aniline, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(4-Chlorophenoxy)aniline |
| CAS | 2770-11-8 |
| Molecular Formula | C12 H10 Cl N O |
| Quantity | 2.5 g |
| Synonym | 2-(4-Chlorophenoxy)benzenamine, o-(p-Chlorophenoxy)aniline, 2-Amino-4'-chlorodiphenyl Ether |
| InChI Formula | InChI=1S/C12H10ClNO/c13-9-5-7-10(8-6-9)15-12-4-2-1-3-11(12)14/h1-8H,14H2 |
| SMILES | Nc1ccccc1Oc2ccc(Cl)cc2 |
| IUPAC Name | 2-(4-chlorophenoxy)aniline |
| Molecular Weight (g/mol) | 219.67 |
| Formula Weight | 219.0451 |