Learn More
2-(4-Bromophenyl)-2,2’-dimethylacetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(4-Bromophenyl)-2,2'-dimethylacetic Acid |
| CAS | 32454-35-6 |
| Molecular Formula | C10 H11 Br O2 |
| Quantity | 2.5 g |
| Synonym | Benzeneacetic acid, 4-bromo-α,α-dimethyl-, Hydratropic acid, p-bromo-α-methyl- (6CI,8CI), 4-Bromo-α,α-dimethylbenzeneacetic acid, 2-(4-Bromophenyl)-2-methylpropanoic acid, 2-(4-Bromophenyl)-2-methylpropionic acid, 2-(4-Bromophenyl)-2,2'-dimethylacetic acid |
| InChI Formula | InChI=1S/C10H11BrO2/c1-10(2,9(12)13)7-3-5-8(11)6-4-7/h3-6H,1-2H3,(H,12,13) |
| SMILES | CC(C)(C(=O)O)c1ccc(Br)cc1 |
| IUPAC Name | 2-(4-bromophenyl)-2-methylpropanoic acid |
| Molecular Weight (g/mol) | 243.1 |
| Formula Weight | 241.9942 |