Learn More
2-(4-Aminophenyl)propanoic Acid, TRC
Product Code. 30328423
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328423 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30328423 Supplier Toronto Research Chemicals Supplier No. TRCA62269010G
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(4-Aminophenyl)propanoic Acid
CAS 59430-62-5
Molecular Formula C9H11NO2
Quantity 10 g
Synonym 2-(4-Aminophenyl)propanoic Acid, 2-(4-Aminophenyl)propionic Acid, 2-(p-Aminophenyl)propionic Acid, 4-Amino-α-methylbenzeneacetic Acid, p-Amino-α-methylphenylacetic Acid, α-(4-Aminophenyl)-α-methylacetic Acid, α-(p-Aminophenyl)propionic Acid
InChI Formula InChI=1S/C9H11NO2/c1-6(9(11)12)7-2-4-8(10)5-3-7/h2-6H,10H2,1H3,(H,11,12)
SMILES CC(C(=O)O)c1ccc(N)cc1
IUPAC Name 2-(4-aminophenyl)propanoic acid
Molecular Weight (g/mol) 165.19
Formula Weight 165.079
Product Title
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.