missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorophenyl isocyanate, tech. 85%

Product Code. 11304276
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304276 1 g 1g
11314276 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304276 Supplier Thermo Scientific Alfa Aesar Supplier No. L19349.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 50528-80-8 | C7H2F3NO | 173.09 g/mol

2,4,6-Trifluorophenyl isocyanate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trifluorophenyl isocyanate is used as a pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, amines and alcohols. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50528-80-8
Molecular Formula C7H2F3NO
Molecular Weight (g/mol) 173.09
MDL Number MFCD03426028
InChI Key XORSGSHLSDFOHP-UHFFFAOYSA-N
Synonym 2,4,6-trifluorophenyl isocyanate, 2,4,6-trifluorophenylisocyanate, benzene, 1,3,5-trifluoro-2-isocyanato, 2,4,6-trifluorobenzenisocyanate, pubchem4459, acmc-209xbb, ksc495i7j, 1,3,5-trifluoro-2-isocyatobenzene, 2,4,6-trifluoro-1-isocyanatobenzene, 1,3,5-trifluoro-2-isocyanato-benzene
PubChem CID 2783237
IUPAC Name 1,3,5-trifluoro-2-isocyanatobenzene
SMILES FC1=CC(F)=C(N=C=O)C(F)=C1

Specifications

Refractive Index 1.476
Quantity 1 g
UN Number UN3080
Beilstein 6325663
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 173.11
Percent Purity 85%
Grade Technical
Chemical Name or Material 2,4,6-Trifluorophenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.