missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzotrifluoride, 98%, Thermo Scientific™

Product Code. 10663702 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10663702 5 g Glass bottle 5g
10546441 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10663702 Supplier Thermo Scientific Acros Supplier No. 347520050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122030-04-0
Molecular Formula C7H2F6
Molecular Weight (g/mol) 200.083
MDL Number MFCD00190109
InChI Key SGPXFVDJRSZIPK-UHFFFAOYSA-N
Synonym 2,4,6-trifluorobenzotrifluoride, 1,3,5-trifluoro-2-trifluoromethyl benzene, pubchem4877, 2,4,6-trifluorobenzotrifluorode, 2,4,6-tri-fluorobenzotrifluoride, benzene, 1,3,5-trifluoro-2-trifluoromethyl, alpha,alpha,alpha,2,4,6-hexafluorotoluene, alpha,alpha,alpha,2,4,6-hexafluorotoluene;1,3,5-trifluoro-2-trifluoromethyl benzene
PubChem CID 2734030
IUPAC Name 1,3,5-trifluoro-2-(trifluoromethyl)benzene
SMILES C1=C(C=C(C(=C1F)C(F)(F)F)F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.3840 to 1.3860 (20°C, 589nm)
Quantity 5 g
Formula Weight 200.08
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2, 4, 6-Trifluorobenzotrifluoride, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.