Learn More
2,4,6-Triacetylphloroglucinol, TRC
Product Code. 30456838
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456838 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30456838 Supplier Toronto Research Chemicals Supplier No. TRCT7670121G
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4,6-Triacetylphloroglucinol
CAS 2161-87-7
Molecular Formula C12 H12 O6
Quantity 1 g
InChI Formula InChI=1S/C12H12O6/c1-4(13)7-10(16)8(5(2)14)12(18)9(6(3)15)11(7)17/h16-18H,1-3H3
SMILES CC(=O)c1c(O)c(C(=O)C)c(O)c(C(=O)C)c1O
IUPAC Name 1-(3,5-diacetyl-2,4,6-trihydroxyphenyl)ethanone
Molecular Weight (g/mol) 252.22
Formula Weight 252.0634