Learn More
2,4,5-Trifluorobenzoic acid, 98%
Product Code. 11494100
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494100 25 g 25g
11442267 1 g 1g
11452267 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11494100 Supplier Thermo Scientific Alfa Aesar Supplier No. A14193.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 446-17-3 | C7H2F3O2 | 175.09 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 446-17-3
Molecular Formula C7H2F3O2
Molecular Weight (g/mol) 175.09
MDL Number MFCD00013306
InChI Key AKAMNXFLKYKFOJ-UHFFFAOYSA-M
Synonym 2,4,5-trifluoro-benzoic acid, benzoic acid, 2,4,5-trifluoro, pubchem1346, acmc-209jyu, 2,4,5-trifluorobenzoicacid, ksc235o3j, 2,4,5-trifluoro benzoic acid, rarechem al bo 0258, akamnxflkykfoj-uhfffaoysa, 2,4,5-trifluorobenzoic acid
PubChem CID 521170
IUPAC Name 2,4,5-trifluorobenzoic acid
SMILES [O-]C(=O)C1=CC(F)=C(F)C=C1F
Specifications
Melting Point 95°C to 100°C
Quantity 25 g
Beilstein 3257609
Formula Weight 176.1
Percent Purity 98%
Chemical Name or Material 2,4,5-Trifluorobenzoic acid