missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4,5-Trichlorobenzenesulfonic acid potassium salt, 90+%

Product Code. 11455566
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455566 1 g 1g
11465566 5 g 5g
11475566 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455566 Supplier Alfa Aesar™ Supplier No. B20713.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 62625-17-6
Molecular Formula C6H2Cl3KO3S
Molecular Weight (g/mol) 299.587
MDL Number MFCD00024894
InChI Key DRPLRPSXUUSFOB-UHFFFAOYSA-M
PubChem CID 23692292
IUPAC Name potassium;2,4,5-trichlorobenzenesulfonate
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)[O-].[K+]

Specifications

Melting Point >300°C
Quantity 1 g
Sensitivity Hygroscopic
Synonym potassium 2,4,5-trichlorobenzenesulfonate, benzenesulfonic acid, 2,4,5-trichloro-, potassium salt, 2,4,5-trichlorobenzenesulfonic acid potassium salt, potassium 2,4,5-trichlorobenzenesulphonate, benzenesulfonic acid, 2,4,5-trichloro-, potassium salt 1:1, potassium 2,4,5-trichlorobenzene-1-sulfonate, 2,4,5-trichlorobenzenesulfonic acid, potassium salt, c6h2cl3so3.k, potassium2,4,5-trichlorobenzenesulfonate, potassium ion 2,4,5-trichlorobenzenesulfonate
Formula Weight 299.6
Percent Purity ≥90%
Chemical Name or Material 2,4,5-Trichlorobenzenesulfonic acid potassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.