Learn More
2,4:3,5-Di-O-methylene-D-gluconic Acid Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4:3,5-Di-O-methylene-D-gluconic Acid Methyl Ester |
| CAS | 27934-92-5 |
| Molecular Formula | C9H14O7 |
| Quantity | 250 mg |
| Synonym | Methyl 2,4:3,5-di-O-methylene-D-gluconate |
| InChI Formula | InChI=1S/C9H14O7/c1-12-9(11)8-7-6(14-4-16-8)5(2-10)13-3-15-7/h5-8,10H,2-4H2,1H3/t5-,6-,7+,8-/m1/s1 |
| SMILES | O=C([C@H](OCO1)[C@@H](OCO2)[C@H]1[C@H]2CO)OC |
| IUPAC Name | methyl (4R,4aS,8R,8aR)-8-(hydroxymethyl)tetrahydro-[1,3]dioxino[5,4-d][1,3]dioxine-4-carboxylate |
| Molecular Weight (g/mol) | 234.2 |
| Formula Weight | 234.07 |