Learn More
2-(4-(2,2-Dichlorocyclopropyl)phenoxy)-2-methylpropan-1-ol, TRC
Product Code. 30368915
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368915 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30368915 Supplier Toronto Research Chemicals Supplier No. TRCD43404510MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(4-(2,2-Dichlorocyclopropyl)phenoxy)-2-methylpropan-1-ol
CAS 2469279-29-4
Molecular Formula C13H16Cl2O2
Quantity 10 mg
Synonym 2-[4-[(1RS)-2,2-Dichlorocyclopropyl]phenoxy]-2-methylpropan-1-ol, Ciprofibrate Imp. C (EP)
InChI Formula InChI=1S/C13H16Cl2O2/c1-12(2,8-16)17-10-5-3-9(4-6-10)11-7-13(11,14)15/h3-6,11,16H,7-8H2,1-2H3
SMILES CC(C)(CO)Oc1ccc(cc1)C2CC2(Cl)Cl
Molecular Weight (g/mol) 275.17