Learn More
2,3-Xylohydroquinone, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3-Dimethylhydroquinone |
| CAS | 608-43-5 |
| Molecular Formula | C8 H10 O2 |
| Quantity | 1 g |
| Synonym | 1,4-Benzenediol','2,3-dimethyl-, Hydroquinone','2,3-dimethyl- (7CI,8CI), 2,3-Dimethyl-1,4-benzenediol, 1,4-Dihydroxy-2,3-dimethylbenzene, 2,3-Dimethyl-p-hydroquinone, 2,3-Dimethylhydroquinone, 2,3-Xylohydroquinone, 4-Hydroxy-2,3-Dimethylphenol, NSC 108080, o-Xylene-3,6-diol, o-Xylohydroquinone, 2,3-Dimethylbenzene-1,4-diol |
| InChI Formula | InChI=1S/C8H10O2/c1-5-6(2)8(10)4-3-7(5)9/h3-4,9-10H,1-2H3 |
| SMILES | Cc1c(C)c(O)ccc1O |
| IUPAC Name | 2,3-dimethylbenzene-1,4-diol |
| Molecular Weight (g/mol) | 138.16 |
| Formula Weight | 138.0681 |