missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Pyridyl)thiazolidine, 97%, Thermo Scientific™

Catalog Number 15428668
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
700-93-6
C8H10N2S
166.24
MFCD01691463
SMYGUEYUDFIZCL-UHFFFAOYNA-N
3-1,3-thiazolidin-2-yl pyridine, 3-2-thiazolidinyl pyridine, pyridine, 3-2-thiazolidinyl, 2-3-pyridyl thiazolidine
120769
2-pyridin-3-yl-1,3-thiazolidine
C1CSC(N1)C1=CC=CN=C1
This item is not returnable. View return policy
700-93-6
C8H10N2S
166.24
MFCD01691463
SMYGUEYUDFIZCL-UHFFFAOYNA-N
3-1,3-thiazolidin-2-yl pyridine, 3-2-thiazolidinyl pyridine, pyridine, 3-2-thiazolidinyl, 2-3-pyridyl thiazolidine
120769
2-pyridin-3-yl-1,3-thiazolidine
C1CSC(N1)C1=CC=CN=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 166.24
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(3-Pyridyl)thiazolidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'