Learn More
2-(3-Phenylpropyl)pyridine, 99%
Product Code. 11450687
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450687 25 g 25g
11430687 1 g 1g
11440687 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11450687 Supplier Thermo Scientific Alfa Aesar Supplier No. B22495.14
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 2110-18-1 | C14H15N | 197.28 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2110-18-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
MDL Number MFCD00191237
InChI Key JJJPNTQYUJPWGQ-UHFFFAOYSA-N
Synonym 2-3-phenylpropyl pyridine, pyridine, 2-3-phenylpropyl, unii-9f0bfm744f, alpha-3-phenylpropyl pyridine, 2-phenylpropylpyridine, cortex pyridine, phenylpropylpyridine, 3-phenylpropyl pyridine, acmc-1cs6o, dsstox_cid_22356
PubChem CID 459494
IUPAC Name 2-(3-phenylpropyl)pyridine
SMILES C(CC1=CC=CC=C1)CC1=CC=CC=N1
Specifications
Density 1.017
Boiling Point 142°C to 143°C (1 mmHg)
Flash Point 100°C (212°F)
Quantity 25 g
Formula Weight 197.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(3-Phenylpropyl)pyridine