Learn More
2',3'-O-Isopropylidene-5-fluorouridine, TRC
Specifications
Specifications
| Chemical Name or Material | 2',3'-O-Isopropylidene-5-fluorouridine |
| CAS | 2797-17-3 |
| Molecular Formula | C12H15FN2O6 |
| Quantity | 1 g |
| Synonym | 5-Fluoro-2',3'-O-(1-methylethylidene)uridine, 5-Fluoro-2',3'-O-isopropylideneuridine, 5-Fluoro-2',3'-isopropylideneuridine |
| InChI Formula | InChI=1S/C12H15FN2O6/c1-12(2)20-7-6(4-16)19-10(8(7)21-12)15-3-5(13)9(17)14-11(15)18/h3,6-8,10,16H,4H2,1-2H3,(H,14,17,18) |
| SMILES | CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N3C=C(F)C(=O)NC3=O |
| Molecular Weight (g/mol) | 302.26 |