Learn More
2-[3-Fluoro-4-(trifluoromethyl)phenyl]-4-methyl-5-hydroxymethyl Thiazole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-[3-Fluoro-4-(trifluoromethyl)phenyl]-4-methyl-5-hydroxymethyl thiazole |
| CAS | 317319-27-0 |
| Molecular Formula | C12 H9 F4 N O S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C12H9F4NOS/c1-6-10(5-18)19-11(17-6)7-2-3-8(9(13)4-7)12(14,15)16/h2-4,18H,5H2,1H3 |
| SMILES | Cc1nc(sc1CO)c2ccc(c(F)c2)C(F)(F)F |
| IUPAC Name | [2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazol-5-yl]methanol |
| Molecular Weight (g/mol) | 291.26 |
| Formula Weight | 291.0341 |