missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylnaphthalene, 97%

Product Code. p-7044823
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11398125

Brand: Thermo Scientific Alfa Aesar L08386.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 581-40-8 | C12H12 | 156.228 g/mol

It is used in the identification of individual aromatic hydrocarbons in kerosene fraction (b.p. 150-250°).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the identification of individual aromatic hydrocarbons in kerosene fraction (b.p. 150-250°).

Solubility
Solubility in toluene (almost transparency). Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 581-40-8
Molecular Formula C12H12
Molecular Weight (g/mol) 156.228
MDL Number MFCD00004119
InChI Key WWGUMAYGTYQSGA-UHFFFAOYSA-N
Synonym guajen, naphthalene, 2,3-dimethyl, unii-35dj6se17o, 2,3-dmn, 2,3-dimethyl naphthalene, acmc-1ao3c, 2,3-dimethylnaphthalene, naphthalene, 2,3-dimethyl-8ci 9ci
PubChem CID 11386
ChEBI CHEBI:48615
IUPAC Name 2,3-dimethylnaphthalene
SMILES CC1=CC2=CC=CC=C2C=C1C

Specifications

Melting Point 103°C to 106°C
Boiling Point 265°C to 268°C
Quantity 1 g
Beilstein 1852621
Formula Weight 156.23
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylnaphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.