Learn More
2,3-Dimethyl-1-butene, 99%
Product Code. p-7044287
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11307545
Brand: Thermo Scientific Alfa Aesar L05675.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 563-78-0 | C6H12 | 84.162 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 563-78-0
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00008923
InChI Key OWWIWYDDISJUMY-UHFFFAOYSA-N
Synonym 2,3-dimethyl-1-butene, 1-butene, 2,3-dimethyl, 2,3-dimethylbutene, 2,3-dimethylbutene-1, 2,3-dimethyl butene, acmc-1aqwm, butene, 2,3-dimethyl, ch3 2chc ch3 =ch2, 1-butene, 2,3-dimethyl-8ci 9ci
PubChem CID 11249
IUPAC Name 2,3-dimethylbut-1-ene
SMILES CC(C)C(=C)C
Specifications
Melting Point -157°C
Boiling Point 56°C to 57°C
Flash Point −18°C (−°F)
Quantity 5 g
UN Number UN3295
Beilstein 505982
Formula Weight 84.16
Percent Purity 99%
Chemical Name or Material 2,3-Dimethyl-1-butene