missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrothieno[3,4-b][1,4]dioxin-5-ylmethanol, ≥97%, Thermo Scientific™

Product Code. p-3175034
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10341673 250 mg Amber Glass Bottle 250mg
10680705 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10341673 Supplier Thermo Scientific Maybridge Supplier No. CC47809CB
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 859851-01-7
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD08060539
InChI Key LRWVQOFWMRDMHM-UHFFFAOYSA-N
Synonym 2,3-dihydrothieno 3,4-b 1,4 dioxin-5-ylmethanol, 2h,3h-thieno 3,4-b 1,4 dioxin-5-ylmethanol, thieno 3,4-b-1,4-dioxin-5-methanol,2,3-dihydro, 3,4-ethylenedioxy-thiophene-methanol, 2-hydroxymethyl-3,4-ethylene dioxythiophene, 2,3-dihydrothieno 3,4-b-1,4-dioxin-5-methanol, 2h,3h-thiopheno 3,4-e 1,4-dioxan-5-ylmethan-1-ol, 2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl methanol, thieno 3,4-b-1,4-dioxin-5-methanol, 2,3-dihydro
PubChem CID 7537657
SMILES OCC1=C2OCCOC2=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 2,3-dihydrothieno[3,4-b][1,4]dioxin-5-ylmethanol
Formula Weight 172.2
Percent Purity 97%
Chemical Name or Material 2,3-Dihydrothieno[3,4-b][1,4]dioxin-5-ylmethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.