Learn More
2,3-Dihydro-5-mercapto-3-oxo-4-isothiazolecarboxylic Acid Trisodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3-Dihydro-5-mercapto-3-oxo-4-isothiazolecarboxylic Acid Trisodium Salt |
| CAS | 76857-14-2 |
| Molecular Formula | C4NNa3O3S2 |
| Quantity | 10 g |
| Synonym | 2,3-Dihydro-5-mercapto-3-oxo-4-isothiazolecarboxylic Acid Sodium Salt (1:3) |
| InChI Formula | InChI=1S/C4H3NO3S2.3Na/c6-2-1(3(7)8)4(9)10-5-2;;;/h9H,(H,5,6)(H,7,8);;;/q;3*+1/p-3 |
| SMILES | [Na+].[Na+].[Na+].[O-]C(=O)c1c([O-])nsc1[S-] |
| IUPAC Name | trisodium;3-oxido-5-sulfido-1,2-thiazole-4-carboxylate |
| Molecular Weight (g/mol) | 243.15 |
| Formula Weight | 242.9013 |