missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-5-benzofuranacetic Acid Methyl Ester, TRC

Product Code. 30384676
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384676 100 mg
30385570 1 g
30387875 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384676 Supplier Toronto Research Chemicals Supplier No. TRCD448495100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155852-41-8
Molecular Formula C11 H12 O3
Molecular Weight (g/mol) 192.21
IUPAC Name methyl 2-(2,3-dihydro-1-benzofuran-5-yl)acetate
SMILES COC(=O)Cc1ccc2OCCc2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H12O3/c1-13-11(12)7-8-2-3-10-9(6-8)4-5-14-10/h2-3,6H,4-5,7H2,1H3
Formula Weight 192.0786
Chemical Name or Material 2,3-Dihydro-5-benzofuranacetic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.