Learn More
2,3-Dihydro-3,5-dihydroxy-6-methyl-4-pyrone, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3-Dihydro-3,5-dihydroxy-6-methyl-4-pyrone |
| CAS | 28564-83-2 |
| Molecular Formula | C6H8O4 |
| Quantity | 100 mg |
| Synonym | 2,3-Dihydro-3,5-dihydroxy-6-methyl-4-pyranone, 3-Hydroxy-2,3-dihydromaltol, 5-Hydroxy-5,6-dihydromaltol, 6-Methyl-3,5-dihydroxy-2,3-dihydro-4H-pyran-4-one, DDMP, Pyranone |
| InChI Formula | InChI=1S/C6H8O4/c1-3-5(8)6(9)4(7)2-10-3/h4,7-8H,2H2,1H3 |
| SMILES | CC1=C(O)C(=O)C(O)CO1 |
| IUPAC Name | 3,5-dihydroxy-6-methyl-2,3-dihydropyran-4-one |
| Molecular Weight (g/mol) | 144.13 |
| Formula Weight | 144.0423 |